2-Benzyloxy-4-fluorphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Benzyloxy)-4-fluorophenyl)boronic acid, 2-Benzyloxy-4-fluorobenzeneboronic acid
PC10105-5G
APOSPC10105-5GEA
58
EUR
APOSPC10105-5G
2-Benzyloxy-4-fluorphenylboronsäure
2-Benzyloxy-4-fluorphenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molecular Weight: 246,04 g/mol Schmelzpunkt: 110…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788402 CAS-Nummer: 848779-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...