2-Benzyloxy-5-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Benzyloxy)-5-fluorophenyl)boronic acid, 2-Benzyloxy-5-fluorobenzeneboronic acid
PC10106-500MG
APOSPC10106-500MGEA
21.9
EUR
APOSPC10106-500MG
2-Benzyloxy-5-fluorphenylboronsäure 98%
2-Benzyloxy-5-fluorphenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molecular Weight: 246,0420 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03788403 CAS-Nummer: 779331-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...