Ethyl-(3,5-bis(trifluormethyl)phenyl)acetat 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(3,5-bis(trifluoromethyl)phenyl)acetate

PC1011-25G PC1011-50G PC1011-5G
APOSPC1011-25GEA 553 EUR
APOSPC1011-25G APOSPC1011-50G APOSPC1011-5G
Ethyl-(3,5-bis(trifluormethyl)phenyl)acetat 97%
Ethyl-(3,5-bis(trifluormethyl)phenyl)acetat
Formel: C₁₂H₁₀F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08689894
CAS-Nummer: 144632-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR