1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid

Lieferant: Apollo Scientific

PC10111-1G PC10111-500MG
APOSPC10111-1GEA 244 EUR
APOSPC10111-1G APOSPC10111-500MG
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
Formel: C₁₂H₁₂ClF₃N₂O₂
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172245
CAS-Nummer: 337919-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR