2,5-Difluor-4-nitrobenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC10116-1G
PC10116-25G
PC10116-5G
APOSPC10116-1GEA
21.9
EUR
APOSPC10116-1G
APOSPC10116-25G
APOSPC10116-5G
2,5-Difluor-4-nitrobenzoesäure 98%
2,5-Difluoro-4-nitrobenzoic acid
Formel:
C₇H₃F₂NO₄ Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425705 CAS-Nummer: 116465-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...