3-Amino-4-fluorobenzoic acid hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1012-1G
PC1012-5G
APOSPC1012-1GEA
178
EUR
APOSPC1012-1G
APOSPC1012-5G
3-Amino-4-fluorobenzoic acid hydrochloride 97%
3-Amino-4-fluorobenzoic acid hydrochloride
Formel:
C₇H₆FNO₂·ClH Schmelzpunkt: 240…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656013 CAS-Nummer: 3799-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...