1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

Lieferant: Apollo Scientific

PC10123-1G PC10123-500MG PC10123-5G
APOSPC10123-1GEA 217 EUR
APOSPC10123-1G APOSPC10123-500MG APOSPC10123-5G
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Formel: C₁₇H₁₃F₄N₃
Schmelzpunkt: 109…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791145
CAS-Nummer: 446275-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR