5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Lieferant: Apollo Scientific
|
Gefahr
|
PC10127-1G
APOSPC10127-1GEA
0
EUR
APOSPC10127-1G
5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
5-(Chloromethyl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Formel:
C₁₀H₆ClF₃N₂O Siedepunkt: 87…90 °C/0,2mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200906 CAS-Nummer: 844498-80-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...