Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate
Lieferant: Apollo Scientific
PC10133-1G
PC10133-500MG
PC10133-5G
APOSPC10133-1GEA
405
EUR
APOSPC10133-1G
APOSPC10133-500MG
APOSPC10133-5G
Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate
Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate
Formel:
C₁₀H₉ClFNO₅ Molecular Weight: 277.64 g/mol Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425715 CAS-Nummer: 439094-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...