Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate

Lieferant: Apollo Scientific

PC10133-1G PC10133-500MG PC10133-5G
APOSPC10133-1GEA 405 EUR
APOSPC10133-1G APOSPC10133-500MG APOSPC10133-5G
Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate
Ethyl-2-(2-chloro-4-fluoro-5-nitrophenoxy)acetate
Formel: C₁₀H₉ClFNO₅
Molecular Weight: 277.64 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425715
CAS-Nummer: 439094-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR