5-Amino-2-chloro-4-fluorophenol
Lieferant: Apollo Scientific
|
Achtung
|
PC10135-1G
APOSPC10135-1GEA
21.9
EUR
APOSPC10135-1G
5-Amino-2-chloro-4-fluorophenol
5-Amino-2-chloro-4-fluorophenol
Formel:
C₆H₅ClFNO Schmelzpunkt: 157…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055622 CAS-Nummer: 84478-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...