Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

PC10137-5G
APOSPC10137-5GEA 69.6 EUR
APOSPC10137-5G
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate
Formel: C₈H₉F₃N₂O₂
Molecular Weight: 222,167 g/mol
Schmelzpunkt: 60…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02083398
CAS-Nummer: 111493-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR