3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol

Lieferant: Apollo Scientific
Achtung

PC10139-1G
APOSPC10139-1GEA 873 EUR
APOSPC10139-1G
3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol
3-[Cyclohexyl(methyl)amino]-1,1,1-trifluoro-2-propanol
Formel: C₁₀H₁₈F₃NO
Molecular Weight: 225,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001225
CAS-Nummer: 446276-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR