4,4'-Di(hydroxy)octafluorobiphenyl hydrate

Lieferant: Apollo Scientific
Achtung

PC1015-250MG PC1015-500MG
APOSPC1015-250MGEA 21.9 EUR
APOSPC1015-250MG APOSPC1015-500MG
4,4'-Di(hydroxy)octafluorobiphenyl hydrate
4,4'-Di(hydroxy)octafluorobiphenyl hydrate
Formel: C₁₂H₄F₈O₃
Schmelzpunkt: 209…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149466
CAS-Nummer: 205926-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR