4-(Trifluormethyl)nicotinamid
Lieferant: Apollo Scientific
|
Achtung
|
PC10153-1G
PC10153-10G
APOSPC10153-1GEA
36.4
EUR
APOSPC10153-1G
APOSPC10153-10G
4-(Trifluormethyl)nicotinamid
4-(Trifluormethyl)nicotinamid
Formel:
C₇H₅F₃N₂O Molecular Weight: 190.1226 g/mol Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126658 CAS-Nummer: 158062-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...