2-(4-Fluorphenoxy)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC10172-1G
APOSPC10172-1GEA
102
EUR
APOSPC10172-1G
2-(4-Fluorphenoxy)benzaldehyd
2-(4-Fluorophenoxy)benzaldehyde
Formel:
C₁₃H₉FO₂ Molecular Weight: 216,21 g/mol Siedepunkt: 120…122 °C/0,05mmHg Schmelzpunkt: 54…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01568770 CAS-Nummer: 320423-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...