N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide
Lieferant: Apollo Scientific
PC10178-1G
PC10178-500MG
PC10178-5G
APOSPC10178-1GEA
217
EUR
APOSPC10178-1G
APOSPC10178-500MG
APOSPC10178-5G
N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide
N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide
Formel:
C₇H₇F₂N₃O Molecular Weight: 187.15 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791164 CAS-Nummer: 171277-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...