Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Lieferant: Apollo Scientific
|
Achtung
|
PC1018-1G
APOSPC1018-1GEA
208
EUR
APOSPC1018-1G
Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Formel:
C₇F₁₅O₅S·K Schmelzpunkt: 250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155781 CAS-Nummer: 70755-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...