2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride

Lieferant: Apollo Scientific

PC10205-500MG
APOSPC10205-500MGEA 688 EUR
APOSPC10205-500MG
2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride
2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride
Formel: C₁₄H₁₅ClF₃N₄·Cl
Molecular Weight: 367,197 g/mol
Schmelzpunkt: 175…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793064
CAS-Nummer: 317822-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR