2-(4-Fluorophenoxy)benzenecarbaldehyde oxime

Lieferant: Apollo Scientific

PC10206-1G PC10206-500MG
APOSPC10206-1GEA 799 EUR
APOSPC10206-1G APOSPC10206-500MG
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
Formel: C₁₃H₁₀FNO₂
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091015
CAS-Nummer: 449778-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR