2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC10209-1G
APOSPC10209-1GEA
488
EUR
APOSPC10209-1G
2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid
2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid
Formel:
C₁₂H₈F₃NO₂S Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02186742 CAS-Nummer: 478030-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...