1-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone
Lieferant: Apollo Scientific
|
Achtung
|
PC10216-1G
APOSPC10216-1GEA
81.2
EUR
APOSPC10216-1G
1-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone
1-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone
Formel:
C₁₅H₁₂ClFO₂ Schmelzpunkt: 60…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001254 CAS-Nummer: 400878-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...