7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-(Diethylamino)-4-(trifluoromethyl)coumarin-3-carboxylic acid

PC10220-1G
APOSPC10220-1GEA 576 EUR
APOSPC10220-1G
7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid
7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid
Formel: C₁₅H₁₄F₃NO₄
Molecular Weight: 329,28 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425755
CAS-Nummer: 333303-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR