3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Lieferant: Apollo Scientific
PC10221-1G
PC10221-5G
APOSPC10221-1GEA
386
EUR
APOSPC10221-1G
APOSPC10221-5G
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel:
C₁₃H₁₂F₃NO₅ Molecular Weight: 319.24 g/mol Schmelzpunkt: 170…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791204 CAS-Nummer: 886360-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...