3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Lieferant: Apollo Scientific

PC10221-1G PC10221-5G
APOSPC10221-1GEA 386 EUR
APOSPC10221-1G APOSPC10221-5G
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-[4-(Methoxycarbonyl)phenyl]-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel: C₁₃H₁₂F₃NO₅
Molecular Weight: 319.24 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791204
CAS-Nummer: 886360-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR