3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Chlorophenyl)-3-(trifluoroacetamido)propanoic acid

PC10222-10G PC10222-1G PC10222-5G
APOSPC10222-10GEA 1350 EUR
APOSPC10222-10G APOSPC10222-1G APOSPC10222-5G
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel: C₁₁H₉ClF₃NO₃
Molecular Weight: 295.65 g/mol
Schmelzpunkt: 162…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791205
CAS-Nummer: 117291-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR