3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Chlorophenyl)-3-(trifluoroacetamido)propanoic acid
PC10222-10G
PC10222-1G
PC10222-5G
APOSPC10222-10GEA
1350
EUR
APOSPC10222-10G
APOSPC10222-1G
APOSPC10222-5G
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel:
C₁₁H₉ClF₃NO₃ Molecular Weight: 295.65 g/mol Schmelzpunkt: 162…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791205 CAS-Nummer: 117291-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...