Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate

Lieferant: Apollo Scientific

PC10223-1G PC10223-500MG PC10223-5G
APOSPC10223-1GEA 533 EUR
APOSPC10223-1G APOSPC10223-500MG APOSPC10223-5G
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Formel: C₁₃H₁₃F₃N₂O₄
Molecular Weight: 318.25 g/mol
Schmelzpunkt: 231…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791206
CAS-Nummer: 886360-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR