Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Lieferant: Apollo Scientific
PC10223-1G
PC10223-500MG
PC10223-5G
APOSPC10223-1GEA
533
EUR
APOSPC10223-1G
APOSPC10223-500MG
APOSPC10223-5G
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Formel:
C₁₃H₁₃F₃N₂O₄ Molecular Weight: 318.25 g/mol Schmelzpunkt: 231…233 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791206 CAS-Nummer: 886360-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...