3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide

Lieferant: Apollo Scientific

PC10226-1G PC10226-500MG PC10226-5G
APOSPC10226-1GEA 405 EUR
APOSPC10226-1G APOSPC10226-500MG APOSPC10226-5G
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
Formel: C₁₁H₁₀ClF₃N₂O₂
Molecular Weight: 294.66 g/mol
Schmelzpunkt: 241…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03012828
CAS-Nummer: 439110-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR