3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
Lieferant: Apollo Scientific
PC10226-1G
PC10226-500MG
PC10226-5G
APOSPC10226-1GEA
405
EUR
APOSPC10226-1G
APOSPC10226-500MG
APOSPC10226-5G
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
Formel:
C₁₁H₁₀ClF₃N₂O₂ Molecular Weight: 294.66 g/mol Schmelzpunkt: 241…243 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03012828 CAS-Nummer: 439110-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...