3-Amino-2-cyano-3-[4-(4-fluorophenyl)piperazin-1-yl]-N-methylprop-2-enethioamide

Lieferant: Apollo Scientific

PC10231-500MG
APOSPC10231-500MGEA 686 EUR
APOSPC10231-500MG
3-Amino-2-cyano-3-[4-(4-fluorophenyl)piperazin-1-yl]-N-methylprop-2-enethioamide
3-Amino-2-cyano-3-[4-(4-fluorophenyl)piperazin-1-yl]-N-methylprop-2-enethioamide
Formel: C₁₅H₁₈FN₅S
Molecular Weight: 319,4 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00243959
CAS-Nummer: 338422-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR