2-(3,4-Difluorophenoxy)benzenecarboxaldehyde oxime

Lieferant: Apollo Scientific

PC10235-500MG
APOSPC10235-500MGEA 688 EUR
APOSPC10235-500MG
2-(3,4-Difluorophenoxy)benzenecarboxaldehyde oxime
2-(3,4-Difluorophenoxy)benzenecarboxaldehyde oxime
Formel: C₁₃H₉F₂NO₂
Molecular Weight: 249.213 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091017
CAS-Nummer: 451485-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR