2-(4-Chloro-3-fluorophenoxy)benzaldehyde oxime

Lieferant: Apollo Scientific

PC10236-500MG
APOSPC10236-500MGEA 688 EUR
APOSPC10236-500MG
2-(4-Chloro-3-fluorophenoxy)benzaldehyde oxime
2-(4-Chloro-3-fluorophenoxy)benzaldehyde oxime
Formel: C₁₃H₉ClFNO₂
Molecular Weight: 265,667 g/mol
Schmelzpunkt: 67…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02082405
CAS-Nummer: 451485-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR