4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one

Lieferant: Apollo Scientific

PC10239-1G
APOSPC10239-1GEA 873 EUR
APOSPC10239-1G
4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one
4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one
Formel: C₁₆H₁₂ClFO₂
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091020
CAS-Nummer: 451485-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR