N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
Lieferant: Apollo Scientific
|
Achtung
|
PC10243-1G
APOSPC10243-1GEA
36.4
EUR
APOSPC10243-1G
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
Formel:
C₁₄H₁₂FNO₂ Schmelzpunkt: 139…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425776 CAS-Nummer: 131179-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...