3-Methoxy-4-{[3-(trifluoromethyl)benzyl]oxy}benzenecarbaldehyde

Lieferant: Apollo Scientific
Achtung

PC10256-1G
APOSPC10256-1GEA 98.6 EUR
APOSPC10256-1G
3-Methoxy-4-{[3-(trifluoromethyl)benzyl]oxy}benzenecarbaldehyde
3-Methoxy-4-{[3-(trifluoromethyl)benzyl]oxy}benzenecarbaldehyde
Formel: C₁₆H₁₃F₃O₃
Molecular Weight: 310,28 g/mol
Schmelzpunkt: 64…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02255725
CAS-Nummer: 380427-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR