3-Amino-3-(3-fluorphenyl)propionsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-3-(3-fluorophenyl)propanoic acid

PC10275-1G
APOSPC10275-1GEA 21.9 EUR
APOSPC10275-1G
3-Amino-3-(3-fluorphenyl)propionsäure
3-Amino-3-(3-fluorphenyl)propionsäure
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,1796 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00764457
CAS-Nummer: 117391-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR