3-(Boc-amino)-3-(3-fluorphenyl)propionsäure

Lieferant: Apollo Scientific

PC10277-1G PC10277-500MG
APOSPC10277-1GEA 134 EUR
APOSPC10277-1G APOSPC10277-500MG
3-(Boc-amino)-3-(3-fluorphenyl)propionsäure
3-(Boc-amino)-3-(3-fluorphenyl)propionsäure
Formel: C₁₄H₁₈FNO₄
Schmelzpunkt: 120…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090701
CAS-Nummer: 284493-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR