Methyl 2-[(3,3,3-trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylate

Lieferant: Apollo Scientific

PC10279-500MG
APOSPC10279-500MGEA 749 EUR
APOSPC10279-500MG
Methyl 2-[(3,3,3-trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylate
Methyl 2-[(3,3,3-trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylate
Formel: C₁₁H₁₁F₃O₃S
Molecular Weight: 280,263 g/mol
Siedepunkt: 158…160 °C (1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03001288
CAS-Nummer: 385383-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR