1-Brom-2,4-difluor-5-nitrobenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Brom-2,4-difluornitrobenzol

PC10283-5G
APOSPC10283-5GEA 21.9 EUR
APOSPC10283-5G
1-Brom-2,4-difluor-5-nitrobenzol 98%
1-Bromo-2,4-difluoro-5-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237,99 g/mol
Siedepunkt: 85 °C/0,4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00129166
CAS-Nummer: 345-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR