2,2,2-Trifluor-N-phenylacetamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC10285-500MG
APOSPC10285-500MGEA
14.7
EUR
APOSPC10285-500MG
2,2,2-Trifluor-N-phenylacetamid 97%
2,2,2-Trifluor-N-phenylacetamid
Formel:
C₈H₆F₃NO Molecular Weight: 189,1346 g/mol Schmelzpunkt: 89…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00196027 CAS-Nummer: 404-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...