5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(tert-Butyl)-3-(trifluoromethyl)-1H-pyrazole
PC1029-100MG
APOSPC1029-100MGEA
21.9
EUR
APOSPC1029-100MG
5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole 97%
5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole
Formel:
C₈H₁₁F₃N₂ Molecular Weight: 192,19 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00729044 CAS-Nummer: 150433-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...