3-(4-Benzhydrylpiperazino)-1,1,1-trifluoropropan-2-ol
Lieferant: Apollo Scientific
PC10290-500MG
APOSPC10290-500MGEA
688
EUR
APOSPC10290-500MG
3-(4-Benzhydrylpiperazino)-1,1,1-trifluoropropan-2-ol
3-(4-Benzhydrylpiperazino)-1,1,1-trifluoropropan-2-ol
Formel:
C₂₀H₂₃F₃N₂O Molecular Weight: 364,405 g/mol Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871731 CAS-Nummer: 453557-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...