2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(2-Hydroxy-3,3,3-trifluoroprop-1-yl)thio]benzoic acid

PC10296-1G PC10296-500MG
APOSPC10296-1GEA 996 EUR
APOSPC10296-1G APOSPC10296-500MG
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid
Formel: C₁₀H₉F₃O₃S
Molecular Weight: 266.24 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663825
CAS-Nummer: 886361-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR