4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((3-Fluorobenzyl)oxy)benzoic acid, 4-(3-Fluorobenzyloxy)benzoic acid

PC10297-1G
APOSPC10297-1GEA 434 EUR
APOSPC10297-1G
4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid
4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid
Formel: C₁₄H₁₁FO₃
Molecular Weight: 246,24 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663826
CAS-Nummer: 405-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR