4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide

Lieferant: Apollo Scientific

Synonyme: 4-(3-Fluorobenzyloxy)benzhydrazide

PC10298-1G
APOSPC10298-1GEA 434 EUR
APOSPC10298-1G
4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide
4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide
Formel: C₁₄H₁₃FN₂O₂
Molecular Weight: 260,27 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663827
CAS-Nummer: 886361-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR