Pentafluorphenyl-2-propansulfonat

Lieferant: Apollo Scientific
Achtung

PC10303-1G
APOSPC10303-1GEA 132 EUR
APOSPC10303-1G
Pentafluorphenyl-2-propansulfonat
Pentafluorphenyl-2-propansulfonat
Formel: C₉H₇F₅O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD05975149
CAS-Nummer: 886361-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR