Pentafluorphenyl-4-brom-2-chlorbenzolsulfonat 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC10305-1G
APOSPC10305-1GEA
134
EUR
APOSPC10305-1G
Pentafluorphenyl-4-brom-2-chlorbenzolsulfonat 95%
Pentafluorphenyl-4-brom-2-chlorbenzolsulfonat
Formel:
C₁₂H₃BrClF₅O₃S Schmelzpunkt: 88…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05975147 CAS-Nummer: 886361-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...