Pentafluorphenyl-4-nitrobenzolsulfonat

Lieferant: Apollo Scientific

Synonyme: Pentafluorophenyl 4-Nitrobenzenesulphonate

PC10307-25G
APOSPC10307-25GEA 157 EUR
APOSPC10307-25G
Pentafluorphenyl-4-nitrobenzolsulfonat
Pentafluorphenyl-4-nitrobenzolsulfonat
Formel: C₁₂H₄F₅NO₅S
Molecular Weight: 369.2209 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975120
CAS-Nummer: 244633-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR